missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride Dihydrate, ACS Reagent, ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Boiling Point | 1560°C |
Melting Point | 962°C |
Color | White |
pH | 5.2 to 8.2 |
Quantity | 500 g |
UN Number | UN1564 |
Formula Weight | 244.26g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'