missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride dihydrate, ACS, Thermo Scientific Chemicals

Product Code. 11371669
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Barium chloride is usually employed as a chemical reagent to precipitate soluble sulfates and also in the purification of the seawater solution. It is practiced in several applications like heat-treating baths, waste water treatment, the production of PVC stabilizers, oil lubricants, fireworks materials, hardening of steel, in the manufacture of pigments and in the manufacture of other barium salts. It is applied to prepare composites to absorb ammonia and to increase the crystallinty of polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Melting Point 113°C-2H2O
Physical Form Crystalline
Quantity 500 g
UN Number UN1564
Merck Index 14,971
Solubility Information Very soluble in water; Soluble in methanol; Insoluble in ethanol,acetone,ethyl acetate.
IUPAC Name barium(2+) dihydrate dichloride
Formula Weight 244.28 (208.25 Anhydrous)
Grade ACS
Density 3.097
Chemical Name or Material Barium chloride, dihydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Barium chloride dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Barium chloride dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.