missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride Dihydrate, Extra Pure, SLR, Fisher Chemical™
Chemical Identifiers
CAS | 10326-27-9 |
---|---|
Molecular Formula | BaCl2H4O2 |
Molecular Weight (g/mol) | 244.26 |
MDL Number | MFCD00149154 |
InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
IUPAC Name | barium(2+) dihydrate dichloride |
SMILES | O.O.[Cl-].[Cl-].[Ba++] |
Description
Barium Chloride Dihydrate, Extra Pure, SLR
Chemical Identifiers
10326-27-9 | |
244.26 | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
O.O.[Cl-].[Cl-].[Ba++] |
BaCl2H4O2 | |
MFCD00149154 | |
barium(2+) dihydrate dichloride |
Specifications
1560°C | |
10326-27-9 | |
5 | |
BaCl2H4O2 | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
barium(2+) dihydrate dichloride | |
244.28g/mol |
962°C | |
White | |
Solid | |
MFCD00149154 | |
O.O.[Cl-].[Cl-].[Ba++] | |
244.26 | |
Plastic powder jar |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Barium chloride dihydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
Hazard Statement
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.