missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride, ultra dry, 99.95% (metals basis), Thermo Scientific™

Product Code. 11322498
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-37-2
Molecular Formula BaCl2
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003445
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
Synonym barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
PubChem CID 25204
ChEBI CHEBI:63317
SMILES [Cl-].[Cl-].[Ba++]

Specifications

Boiling Point 1560°C
Melting Point 963°C
Color White
Physical Form Beads
Quantity 25 g
UN Number UN1564
Merck Index 14,971
Solubility Information Soluble in water 370g/L.
IUPAC Name barium(2+);dichloride
Formula Weight 208.25
Percent Purity 99.95%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 3.9 g/mL
Assay (metals basis)
Chemical Name or Material Barium chloride, Ultra dry
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.