Learn More
Barium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
193.97 €
Chemical Identifiers
CAS | 10361-37-2 |
---|---|
Molecular Formula | BaCl2 |
Molecular Weight (g/mol) | 208.23 |
MDL Number | MFCD00003445 |
InChI Key | WDIHJSXYQDMJHN-UHFFFAOYSA-L |
Synonym | barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508 |
PubChem CID | 25204 |
ChEBI | CHEBI:63317 |
IUPAC Name | barium(2+);dichloride |
SMILES | [Cl-].[Cl-].[Ba++] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10361-37-2 | |
208.23 | |
WDIHJSXYQDMJHN-UHFFFAOYSA-L | |
25204 | |
barium(2+);dichloride |
BaCl2 | |
MFCD00003445 | |
barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508 | |
CHEBI:63317 | |
[Cl-].[Cl-].[Ba++] |
Specifications
10361-37-2 | |
Beads | |
BaCl2 | |
barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508 | |
[Cl-].[Cl-].[Ba++] | |
208.23 | |
CHEBI:63317 | |
Odorless |
White | |
99.99% (Metals basis) | |
MFCD00003445 | |
WDIHJSXYQDMJHN-UHFFFAOYSA-L | |
barium(2+);dichloride | |
25204 | |
208.25 | |
Barium chloride |
Safety and Handling
H301-H331: Toxic if swallowed. Toxic if inhaled.
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 233-788-1
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.