Learn More
Barium chromate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
65.12 € - 274.37 €
Chemical Identifiers
CAS | 10294-40-3 |
---|---|
Molecular Formula | BaCrO4 |
Molecular Weight (g/mol) | 253.32 |
MDL Number | MFCD00014180 |
InChI Key | QFFVPLLCYGOFPU-UHFFFAOYSA-N |
Synonym | barium chromate, lemon chrome, permanent yellow, barium chromate vi, baryta yellow, ultramarine yellow, pigment yellow 31, ci pigment ylllow 31, barium chromate 1:1, unii-7d7o9cf0ix |
PubChem CID | 25136 |
IUPAC Name | barium(2+) dioxochromiumbis(olate) |
SMILES | [Ba++].[O-][Cr]([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11306148
|
Thermo Scientific Alfa Aesar
011124.06 |
5 g |
65.12 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11316148
|
Thermo Scientific Alfa Aesar
011124.14 |
25 g |
274.37 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium chromate has been found to be useful in many capacities. often the compound is used as a carrier for the chromium ions. is an oxidizing agent, making it useful as a burn rate modifier in pyrotechnic compositions. It is especially useful in delay compositions such as delay fuses. It is also used in the composition of a catalyst for alkane dehydration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10294-40-3 | |
253.32 | |
QFFVPLLCYGOFPU-UHFFFAOYSA-N | |
25136 | |
[Ba++].[O-][Cr]([O-])(=O)=O |
BaCrO4 | |
MFCD00014180 | |
barium chromate, lemon chrome, permanent yellow, barium chromate vi, baryta yellow, ultramarine yellow, pigment yellow 31, ci pigment ylllow 31, barium chromate 1:1, unii-7d7o9cf0ix | |
barium(2+) dioxochromiumbis(olate) |
Specifications
10294-40-3 | |
Powder | |
MFCD00014180 | |
14,972 | |
Insoluble in water. Soluble in strong acids. | |
[Ba++].[O-][Cr]([O-])(=O)=O | |
253.32 | |
253.33 | |
Puratronic™ | |
4.50 g/mL | |
Barium chromate |
Yellow | |
BaCrO4 | |
UN1479 | |
barium chromate, lemon chrome, permanent yellow, barium chromate vi, baryta yellow, ultramarine yellow, pigment yellow 31, ci pigment ylllow 31, barium chromate 1:1, unii-7d7o9cf0ix | |
QFFVPLLCYGOFPU-UHFFFAOYSA-N | |
barium(2+) dioxochromiumbis(olate) | |
25136 | |
99.998% | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P210-P220-P221-P261-P264b-P270-P271-P281-P285-P301+P312-P304+P340-P308+P313-P330-P342+P311-P370+P378q-P501c
H272-H302+H332-H334-H350
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
missing translation for 'einecsNumber' : 233-660-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.