Learn More
Barium fluoride, Optical Grade, Thermo Scientific Chemicals
42.91 € - 292.46 €
Chemical Identifiers
CAS | 7787-32-8 |
---|---|
Molecular Formula | BaF2 |
Molecular Weight (g/mol) | 175.32 |
MDL Number | MFCD00003450 |
InChI Key | OYLGJCQECKOTOL-UHFFFAOYSA-L |
Synonym | barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis |
PubChem CID | 62670 |
SMILES | [F-].[F-].[Ba++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307677
|
Thermo Scientific Alfa Aesar
014057.06 |
5 g |
42.91 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387667
|
Thermo Scientific Alfa Aesar
014057.14 |
25 g |
100.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397667
|
Thermo Scientific Alfa Aesar
014057.22 |
100 g |
292.46 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium fluoride is used in ceramic flux, carbon brushes for electrical equipment, glass making, in the manufacture of other fluorides, crystals for spectroscopy, electronics and dry-film lubricants. Used for the antiseptic. For industry application, such as electron, measuring applicance, metallurgy, ceramics. Barium fluoride is often suitable for applications in the passive IR band (8 to 14 ?m) and is often used as a viewport window for thermography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7787-32-8 | |
175.32 | |
OYLGJCQECKOTOL-UHFFFAOYSA-L | |
62670 |
BaF2 | |
MFCD00003450 | |
barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis | |
[F-].[F-].[Ba++] |
Specifications
2260°C | |
7787-32-8 | |
BaF2 | |
UN1564 | |
barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis | |
OYLGJCQECKOTOL-UHFFFAOYSA-L | |
barium(2+);difluoride | |
62670 | |
Optical | |
1.4741 | |
Barium fluoride |
1353°C | |
Broken Crystals | |
MFCD00003450 | |
14,974 | |
Soluble in methanol,ethanol,water 1.2g/L. | |
[F-].[F-].[Ba++] | |
175.32 | |
175.36 | |
Odorless | |
4.83 g/mL |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
H301-H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'einecsNumber' : 232-108-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only