missing translation for 'onlineSavingsMsg'
Learn More

Barium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 15485955
Click to view available options
Quantity:
2.5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Barium fluoride is used in windows for thermal imaging inspection application in electric power facilities. Barium fluoride can be used to make optical components like lenses, as it is transparent in UV (ultraviolet) and IR (infrared) regions. It is used in windows for IR fuel oil analysis. Barium fluoride is one of the fastest scintillators for the detection of X-rays, and gamma rays. It has been found to be useful for the detection of gamma photons in PET (positron emission tomography), high energy neutrons, alpha and beta particles. Its resistant to high energy radiation is higher than all other optical fluorides. It is employed in metallurgy for refining aluminum, production of welding agents, as a preopacifying agent and in glazing frits production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-32-8
Molecular Formula BaF2
Molecular Weight (g/mol) 175.32
MDL Number MFCD00003450
InChI Key OYLGJCQECKOTOL-UHFFFAOYSA-L
Synonym barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
PubChem CID 62670
SMILES [F-].[F-].[Ba++]

Specifications

Boiling Point 2260°C
Melting Point 1353°C
Physical Form Powder
Quantity 25 g
UN Number UN1564
Merck Index 14,974
Solubility Information Slightly soluble in water; soluble in methanol,and ethanol.
IUPAC Name barium(2+);difluoride
Formula Weight 175.36
Percent Purity 99.99%
Grade Puratronic™
Odor Odorless
Refractive Index 1.4741
Density 4.83 g/mL
Assay (metals basis)
Chemical Name or Material Barium fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'