missing translation for 'onlineSavingsMsg'
Learn More

Barium hexafluoro-2,4-pentanedionate, Thermo Scientific™

Product Code. 11322458
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118131-57-0
Molecular Formula C10H2BaF12O4
Molecular Weight (g/mol) 551.43
MDL Number MFCD00075633 MFCD00075633
InChI Key WUXISMDFUAVWSS-PAMPIZDHSA-L
Synonym barium 2+ 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate, barium 2+ ion 2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
PubChem CID 44717251
IUPAC Name barium(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate)
SMILES [Ba++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 220°C (decomposition)
Physical Form Powder
Quantity 1 g
UN Number UN1564
Formula Weight 551.44
Chemical Name or Material Barium hexafluoro-2,4-pentanedionate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'