Learn More
Barium hydroxide monohydrate, 95%, Thermo Scientific Chemicals
36.00 € - 80.10 €
Chemical Identifiers
CAS | 22326-55-2 |
---|---|
Molecular Formula | BaH4O3 |
Molecular Weight (g/mol) | 189.356 |
MDL Number | MFCD00149151 |
InChI Key | GKQTUHKAQKWLIN-UHFFFAOYSA-L |
Synonym | barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t |
PubChem CID | 16211219 |
IUPAC Name | barium(2+);dihydroxide;hydrate |
SMILES | O.[OH-].[OH-].[Ba+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15412537
|
Thermo Scientific Alfa Aesar
045888.22 |
100 g |
36.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15422537
|
Thermo Scientific Alfa Aesar
045888.36 |
500 g |
80.10 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium hydroxide monohydrate is used in water purification. It is used to prepare lubricating and oil additives as well as a precursor to other barium compounds. It is also used to dehydrate and remove sulfate from various products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22326-55-2 | |
189.356 | |
GKQTUHKAQKWLIN-UHFFFAOYSA-L | |
16211219 | |
O.[OH-].[OH-].[Ba+2] |
BaH4O3 | |
MFCD00149151 | |
barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t | |
barium(2+);dihydroxide;hydrate |
Specifications
22326-55-2 | |
Crystalline | |
Ba(OH)2·H2O | |
UN3262 | |
Soluble in water. | |
O.[OH-].[OH-].[Ba+2] | |
189.356 | |
189.38 (171.36 Anhydrous) | |
Odorless | |
3.743 g/mL |
White | |
BaH4O3 | |
MFCD00149151 | |
barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t | |
GKQTUHKAQKWLIN-UHFFFAOYSA-L | |
barium(2+);dihydroxide;hydrate | |
16211219 | |
95% | |
Air sensitive; Hygroscopic | |
Barium hydroxide monohydrate |
Safety and Handling
GHS H Statement
H301-H314-H318-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H332-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 241-234-5
missing translation for 'rtecsNumber' : CQ9200000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only