missing translation for 'onlineSavingsMsg'
Learn More

Barium Hydroxide Monohydrate, purum p.a., ≥98.0% (T), Honeywell™

Purum p.a.,≥98.0% (T)

55.53 € - 57.57 €

Chemical Identifiers

CAS 22326-55-2
Molecular Formula BaH4O3
Molecular Weight (g/mol) 189.356
MDL Number MFCD00149151
InChI Key GKQTUHKAQKWLIN-UHFFFAOYSA-L
Synonym barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t
PubChem CID 16211219
IUPAC Name barium(2+);dihydroxide;hydrate
SMILES O.[OH-].[OH-].[Ba+2]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15621080
View Documents
Honeywell Research Chemicals
11783-100G
100 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15631080
View Documents
Honeywell Research Chemicals
11783-1KG
1 kg Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications

Chemical Identifiers

22326-55-2
189.356
GKQTUHKAQKWLIN-UHFFFAOYSA-L
16211219
O.[OH-].[OH-].[Ba+2]
BaH4O3
MFCD00149151
barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t
barium(2+);dihydroxide;hydrate

Specifications

780°C
White
BaH4O3
MFCD00149151
barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t
O.[OH-].[OH-].[Ba+2]
189.356
189.36g/mol
purum p.a.
300 hPa at 78°C
Barium Hydroxide Monohydrate
22326-55-2
13.1 (21.0g/L at 25°C)
Ba(OH)2 · H2O
UN3262
GKQTUHKAQKWLIN-UHFFFAOYSA-L
barium(2+);dihydroxide;hydrate
16211219
≥98.0% (T)
Powder
3.743g/mL at 25°C
Videos
Safety and Handling

Safety and Handling

P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H302 + H332-H314

missing translation for 'dotInformation' : 8/PGIII

missing translation for 'einecsNumber' : 241-234-5

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.