missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide octahydrate, 97%, Thermo Scientific Chemicals

Product Code. 11430643
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Alfa Aesar Barium hydroxide octahydrate, 97%, Quantity: 500g, CAS: 12230-71-6, Melting Point: 78 deg.C, Molecular Formula: BaH18O10, Molecular Weight: 315.461, Percent Purity: 97%

Barium hydroxide octahydrate has been used in the centromeric heterochromatin banding technnique and to synthesize BaTiO3. The form Barium hydroxide was reported to catalyze the rate of β elimination of phosphoserine residues. It is also used as a general purpose additive for lubricants and greases, sugar fabrication, manufacturing soaps, fat saponification and chemical synthesis of other barium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium hydroxide octahydrate has been used in the centromeric heterochromatin banding technnique and to synthesize BaTiO3. The form Barium hydroxide was reported to catalyze the rate of β elimination of phosphoserine residues. It is also used as a general purpose additive for lubricants and greases, sugar fabrication, manufacturing soaps, fat saponification and chemical synthesis of other barium compounds.

Solubility
Soluble in water (72 mg/ml at 20°C), methanol, ethanol (slightly), and water (56 mg/ml at 15°C). Insoluble in acetone (practically)

Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

Melting Point 78°C
Quantity 500 g
Linear Formula Ba(OH)2·8H2O
UN Number UN3262
Merck Index 14,977
Solubility Information Soluble in water (72mg/ml at 20°C),methanol,ethanol (slightly),and water (56mg/ml at 15°C). Insoluble in acetone (practically)
IUPAC Name barium(2+);dihydroxide;octahydrate
Formula Weight 315.48 (171.36 Anhydrous)
Percent Purity 97%
Odor Odorless
Sensitivity Air Sensitive
Density 2.18
Chemical Name or Material Barium hydroxide octahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Barium hydroxide octahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : barium hydroxide octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.