missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide octahydrate, 98+%, for analysis, Thermo Scientific Chemicals

Product Code. 10286830
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Barium hydroxide octahydrate, 98+%, for analysis, ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, CAS: 12230-71-6, White, Grade: Analysis, Melting Point: 78.0 deg.C, Molecular Formula: H2BaO2.8H2O, Molecular Weight: 315.48, Percent Purity: 98+%

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

Melting Point 78.0°C
Color White
Physical Form Crystals, Crystalline Flakes or Powder
Quantity 250 g
Assay Percent Range 98+%
Linear Formula Ba(OH)2·8H2O
Merck Index 15, 973
Solubility Information Solubility in water: 56g/L (15°C). Other solubilities: freely soluble in methanol, slightly soluble in ethanol, practically insoluble in acetone
IUPAC Name barium(2+) octahydrate dihydroxide
Formula Weight 315.48
Percent Purity ≥98%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 2.18
Density 2.1800g/mL
Chemical Name or Material Barium hydroxide octahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Barium hydroxide octahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.