Learn More
Barium hydroxide octahydrate, 99.95%, (trace metal basis), Thermo Scientific Chemicals
50.10 € - 166.00 €
Chemical Identifiers
CAS | 12230-71-6 |
---|---|
Molecular Formula | BaH18O10 |
Molecular Weight (g/mol) | 315.46 |
MDL Number | MFCD00149152 |
InChI Key | ZUDYPQRUOYEARG-UHFFFAOYSA-L |
Synonym | barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate |
PubChem CID | 17749109 |
IUPAC Name | barium(2+);dihydroxide;octahydrate |
SMILES | O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10492083
|
Thermo Scientific Acros
377820050 |
5 g | Glass bottle |
50.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10198052
|
Thermo Scientific Acros
377820250 |
25 g | Glass bottle |
166.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
12230-71-6 | |
315.46 | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
17749109 | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
BaH18O10 | |
MFCD00149152 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate | |
barium(2+);dihydroxide;octahydrate |
Specifications
78.0°C | |
White | |
99.95% | |
Ba(OH)2·8H2O | |
15, 973 | |
Solubility in water: 56g/L (15°C). Other solubilities: freely soluble in methanol, slightly soluble in ethanol, practically insoluble in acetone | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] | |
315.46 | |
315.48 | |
Trace Metal Basis | |
2.1800g/mL |
12230-71-6 | |
Powder, Crystals and/or Chunks | |
BaH18O10 | |
MFCD00149152 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
barium(2+);dihydroxide;octahydrate | |
17749109 | |
99.95% | |
2.18 | |
Barium hydroxide octahydrate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
- Barium hydroxide octahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only