missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium iodide hydrate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Useful in pyrotechnic devices. A reagent for facile synthesis of poly(phenylcarbyne).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 95% |
Solubility Information | Freely soluble in water. Soluble in alcohol and acetone |
Formula Weight | 391.15 (Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic; Light sensitive |
Chemical Name or Material | Barium iodide hydrate |
Safety and Handling

Product Identifier
- Barium iodide
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Barium iodide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction