Learn More
Barium iodide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals
Light Sensitive and Hygroscopic
Chemical Identifiers
CAS | 13718-50-8 |
---|---|
Molecular Formula | BaI2 |
Molecular Weight (g/mol) | 391.14 |
MDL Number | MFCD00003452 |
InChI Key | SGUXGJPBTNFBAD-UHFFFAOYSA-L |
IUPAC Name | barium(2+) diiodide |
SMILES | [I-].[I-].[Ba++] |
Description
Barium iodide is used to prepare organobarium compounds by reacting with alkyl potassium compounds. It is also involved in the preparation of active barium metal by reducing with lithium biphenyl.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13718-50-8 | |
391.14 | |
SGUXGJPBTNFBAD-UHFFFAOYSA-L | |
[I-].[I-].[Ba++] |
BaI2 | |
MFCD00003452 | |
barium(2+) diiodide |
Specifications
740°C | |
White | |
99.995% (Metals basis) | |
MFCD00003452 | |
SGUXGJPBTNFBAD-UHFFFAOYSA-L | |
barium(2+) diiodide | |
391.15 | |
Hygroscopic; Light sensitive | |
Barium iodide |
13718-50-8 | |
Beads | |
BaI2 | |
Soluble in water,ethanol and acetone. | |
[I-].[I-].[Ba++] | |
391.14 | |
Odorless | |
5.15 |
Safety and Handling
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 237-276-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only