missing translation for 'onlineSavingsMsg'
Learn More

Barium isopropoxide, Thermo Scientific Chemicals

Product Code. 11352478
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24363-37-9
Molecular Formula C6H14BaO2
Molecular Weight (g/mol) 255.503
MDL Number MFCD00050486
InChI Key CPUJSIVIXCTVEI-UHFFFAOYSA-N
Synonym barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate
PubChem CID 6101105
IUPAC Name barium(2+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].[Ba+2]

Specifications

Melting Point ∼200°C (decomposition)
Color Brown to Yellow
Physical Form Powder
Quantity 1 g
Linear Formula Ba[OCH(CH3)2]2
UN Number UN3205
Formula Weight 255.52
Sensitivity Moisture Sensitive
Chemical Name or Material Barium isopropoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.