Learn More
Barium manganate, tech. 90%, Thermo Scientific Chemicals
24.62 € - 75.68 €
Chemical Identifiers
CAS | 7787-35-1 |
---|---|
Molecular Formula | BaMnO4 |
Molecular Weight (g/mol) | 256.26 |
MDL Number | MFCD00011514 |
InChI Key | ZZCNKSMCIZCVDR-UHFFFAOYSA-N |
Synonym | barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese |
PubChem CID | 3084030 |
IUPAC Name | barium(2+);dioxido(dioxo)manganese |
SMILES | [Ba++].[O-][Mn]([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397946
|
Thermo Scientific Alfa Aesar
L13172.06 |
5 g |
24.62 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307956
|
Thermo Scientific Alfa Aesar
L13172.14 |
25 g |
75.68 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7787-35-1 | |
256.26 | |
ZZCNKSMCIZCVDR-UHFFFAOYSA-N | |
3084030 | |
[Ba++].[O-][Mn]([O-])(=O)=O |
BaMnO4 | |
MFCD00011514 | |
barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese | |
barium(2+);dioxido(dioxo)manganese |
Specifications
7787-35-1 | |
MFCD00011514 | |
14,981 | |
ZZCNKSMCIZCVDR-UHFFFAOYSA-N | |
barium(2+);dioxido(dioxo)manganese | |
3084030 | |
90% | |
Moisture sensitive | |
Barium manganate |
BaMnO4 | |
UN3087 | |
barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese | |
[Ba++].[O-][Mn]([O-])(=O)=O | |
256.26 | |
256.28 | |
Technical | |
4.85 |
Safety and Handling
GHS H Statement
H272-H331-H302
May intensify fire; oxidizer.
Toxic if inhaled.
Harmful if swallowed.
P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
H272-H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 232-109-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.