Learn More
Barium niobium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
67.74 € - 264.32 €
Chemical Identifiers
CAS | 12009-14-2 |
---|---|
Molecular Formula | BaNb2O6 |
Molecular Weight (g/mol) | 419.13 |
MDL Number | MFCD00064751 |
InChI Key | FKKSQMDUUDBSGH-UHFFFAOYSA-N |
Synonym | barium niobate, barium niobium oxide, barium 2+ ; oxido dioxo niobium, barium-niobium-oxide, barium 2+ diniobiumoylolate |
PubChem CID | 6093852 |
IUPAC Name | barium(2+) diniobiumoylolate |
SMILES | [Ba++].[O-][Nb](=O)=O.[O-][Nb](=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11313518
|
Thermo Scientific Alfa Aesar
039130.09 |
10 g |
67.74 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11303518
|
Thermo Scientific Alfa Aesar
039130.18 |
50 g |
264.32 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium niobium oxide is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12009-14-2 | |
419.13 | |
FKKSQMDUUDBSGH-UHFFFAOYSA-N | |
6093852 | |
[Ba++].[O-][Nb](=O)=O.[O-][Nb](=O)=O |
BaNb2O6 | |
MFCD00064751 | |
barium niobate, barium niobium oxide, barium 2+ ; oxido dioxo niobium, barium-niobium-oxide, barium 2+ diniobiumoylolate | |
barium(2+) diniobiumoylolate |
Specifications
12009-14-2 | |
BaNb2O6 | |
UN1564 | |
Insoluble in water | |
[Ba++].[O-][Nb](=O)=O.[O-][Nb](=O)=O | |
419.13 | |
419.11 | |
(metals basis) |
Powder | |
MFCD00064751 | |
barium niobate, barium niobium oxide, barium 2+ ; oxido dioxo niobium, barium-niobium-oxide, barium 2+ diniobiumoylolate | |
FKKSQMDUUDBSGH-UHFFFAOYSA-N | |
barium(2+) diniobiumoylolate | |
6093852 | |
99.9% | |
Barium niobium oxide |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'einecsNumber' : 234-543-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only