missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium nitrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2500 g
Description
Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Moisture sensitive. Incompatible with metal,alloy powders, combustible materials, reducing agents, acids, and acid anhydrides.
Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Moisture sensitive. Incompatible with metal,alloy powders, combustible materials, reducing agents, acids, and acid anhydrides.
Specifications
Melting Point | 592°C (decomposition) |
Quantity | 500 g |
Linear Formula | Ba(NO3)2 |
UN Number | UN1446 |
Merck Index | 14,983 |
Solubility Information | Soluble in water. Insoluble in alcohol. |
Formula Weight | 261.35 |
Percent Purity | 99% |
Sensitivity | Hygroscopic |
Density | 3.24 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'