missing translation for 'onlineSavingsMsg'
Learn More

Barium nitrate, ACS, 99+%, Thermo Scientific Chemicals

26.73 € - 165.83 €

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11386588
SDS View Documents
Thermo Scientific Alfa Aesar
012194.18
50 g
26.73 €
50g
Please to purchase this item. Need a web account? Register with us today!
11376588
SDS View Documents
Thermo Scientific Alfa Aesar
012194.36
500 g
53.37 €
500g
Please to purchase this item. Need a web account? Register with us today!
11366588
SDS View Documents
Thermo Scientific Alfa Aesar
012194.A3
2 kg
165.83 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P270-P271-P280-P301+P310-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c

H272-H301-H319-H332

missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: II

missing translation for 'einecsNumber' : 233-020-5

missing translation for 'rtecsNumber' : CQ9625000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium nitrate, ACS, 99+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.