missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Nitrate, Certified AR for Analysis, Fisher Chemical™
Chemical Identifiers
CAS | 10022-31-8 |
---|---|
Molecular Formula | BaN2O6 |
Molecular Weight (g/mol) | 261.34 |
MDL Number | MFCD00003442 |
InChI Key | IWOUKMZUPDVPGQ-UHFFFAOYSA-N |
IUPAC Name | barium(2+) dinitrate |
SMILES | [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Description
Chemical Identifiers
10022-31-8 | |
261.34 | |
IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
BaN2O6 | |
MFCD00003442 | |
barium(2+) dinitrate |
Specifications
592°C | |
White | |
Solid | |
MFCD00003442 | |
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
261.34 | |
Plastic powder jar |
10022-31-8 | |
5.0 to 8.0 | |
BaN2O6 | |
IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
barium(2+) dinitrate | |
261.34g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Barium nitrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.