missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium nitrate, Puriss. p.a., ACS Reagent, ≥99%, ≥52% Ba basis, Honeywell Fluka™
Click to view available options
Quantity:
500 g
5 kg
6 x 500 g
25 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 5 kg |
Linear Formula | Ba(NO3)2 |
UN Number | 1446 |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2] |
Formula Weight | 261.34g/mol |
Percent Purity | ≥99% |
Grade | puriss. p.a. |
Packaging | Plastic Bottle |
Chemical Name or Material | Barium Nitrate, ACS reagent |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'