missing translation for 'onlineSavingsMsg'
Learn More

Barium oxalate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

61.61 € - 647.22 €

Chemical Identifiers

CAS 516-02-9
Molecular Formula C2BaO4
Molecular Weight (g/mol) 225.345
MDL Number MFCD00054429
InChI Key GXUARMXARIJAFV-UHFFFAOYSA-L
Synonym barium oxalate, unii-54r8vvf8zk, ethanedioic acid, barium salt 1:1, barium 2+ ; oxalate, barium 2+ ion oxalate, 54r8vvf8zk, oxalic acid barium, barium 2+ oxalate ion, acmc-1b1e7, ethanedioic acid,barium salt 1:1
PubChem CID 68201
IUPAC Name barium(2+);oxalate
SMILES C(=O)(C(=O)[O-])[O-].[Ba+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11352108
View Documents
Thermo Scientific Alfa Aesar
013009.06
5 g
61.61 €
5g
Please to purchase this item. Need a web account? Register with us today!
11362108
View Documents
Thermo Scientific Alfa Aesar
013009.14
25 g
217.08 €
25g
Please to purchase this item. Need a web account? Register with us today!
11342108
View Documents
Thermo Scientific Alfa Aesar
013009.22
100 g
647.22 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium oxalate, Puratronic isused as a green pyrotechnic colorant generally in specialized pyrotechnic compositions containing magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

516-02-9
225.345
GXUARMXARIJAFV-UHFFFAOYSA-L
68201
C(=O)(C(=O)[O-])[O-].[Ba+2]
C2BaO4
MFCD00054429
barium oxalate, unii-54r8vvf8zk, ethanedioic acid, barium salt 1:1, barium 2+ ; oxalate, barium 2+ ion oxalate, 54r8vvf8zk, oxalic acid barium, barium 2+ oxalate ion, acmc-1b1e7, ethanedioic acid,barium salt 1:1
barium(2+);oxalate

Specifications

400°C (decomposition)
White
C2BaO4
MFCD00054429
14,985
Soluble in water.
C(=O)(C(=O)[O-])[O-].[Ba+2]
225.345
225.36
Puratronic™
2.658 g/mL
Barium oxalate
516-02-9
Powder
BaC2O4
UN1564
barium oxalate, unii-54r8vvf8zk, ethanedioic acid, barium salt 1:1, barium 2+ ; oxalate, barium 2+ ion oxalate, 54r8vvf8zk, oxalic acid barium, barium 2+ oxalate ion, acmc-1b1e7, ethanedioic acid,barium salt 1:1
GXUARMXARIJAFV-UHFFFAOYSA-L
barium(2+);oxalate
68201
99.999%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

missing translation for 'einecsNumber' : 208-216-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.