Learn More
Barium oxide, 99.5% (metals basis excluding Sr), Sr <500ppm, Thermo Scientific Chemicals
Description
Barium oxide is used in cathode ray tubes, crown glass, and catalysts. It is used for the adsorption NO and NO2. It also plays an important role for NO2 storage and reduction. It acts as an efficient ethoxylation catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 1923°C |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN1884 |
Merck Index | 14,986 |
Solubility Information | Sparingly soluble in water. Soluble in ethanol,dilute mineral acids and alkali. |
Formula Weight | 153.33 |
Percent Purity | 99.5% |
Refractive Index | 1.98 |
Density | 5.7 g/mL |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.