missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium perchlorate, anhydrous, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Description
Barium perchlorate, anhydrous is used to make explosives. It is also used as desiccant, ribonuclease determination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless |
Physical Form | Powder |
Quantity | 100 g |
Solubility Information | Soluble in water. |
Formula Weight | 336.24 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 3.20 g/cm3 |
Chemical Name or Material | Barium perchlorate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'