Learn More
Barium perchlorate trihydrate, Reagent Grade, Thermo Scientific Chemicals
64.20 € - 182.00 €
Chemical Identifiers
CAS | 10294-39-0 |
---|---|
Molecular Formula | BaCl2H6O11 |
Molecular Weight (g/mol) | 390.264 |
MDL Number | MFCD00149148 |
InChI Key | NBHDMFAHVJWOHD-UHFFFAOYSA-L |
Synonym | barium perchlorate trihydrate, barium perchlorate trihydrate, reagent grade, ba.2clo4.3h2o, acmc-1bt65, barium perchlorate-water 1/2/3, barium 2+ trihydrate diperchlorate |
PubChem CID | 53249218 |
IUPAC Name | barium(2+);diperchlorate;trihydrate |
SMILES | O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341768
|
Thermo Scientific Alfa Aesar
011656.30 |
250 g |
64.20 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11351768
|
Thermo Scientific Alfa Aesar
011656.A1 |
1 kg |
182.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium perchlorate, trihydrate, reagent is an extremely powerful oxidizing agent, and therefore commonly used in industry and pyrotechnics. It is also used in RPE-For analysis. It is used as an oxidizing agent, for gas drying, for ribonuclease determination and in the water analysis for sulfate titration with Thorin as indicator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10294-39-0 | |
390.264 | |
NBHDMFAHVJWOHD-UHFFFAOYSA-L | |
53249218 | |
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2] |
BaCl2H6O11 | |
MFCD00149148 | |
barium perchlorate trihydrate, barium perchlorate trihydrate, reagent grade, ba.2clo4.3h2o, acmc-1bt65, barium perchlorate-water 1/2/3, barium 2+ trihydrate diperchlorate | |
barium(2+);diperchlorate;trihydrate |
Specifications
10294-39-0 | |
BaCl2H6O11 | |
UN1447 | |
barium perchlorate trihydrate, barium perchlorate trihydrate, reagent grade, ba.2clo4.3h2o, acmc-1bt65, barium perchlorate-water 1/2/3, barium 2+ trihydrate diperchlorate | |
NBHDMFAHVJWOHD-UHFFFAOYSA-L | |
barium(2+);diperchlorate;trihydrate | |
53249218 | |
Reagent | |
2.74 g/mL |
Crystalline | |
MFCD00149148 | |
14,987 | |
Soluble in water, methanol; Slightly soluble in ethanol, ethyl acetate, acetone; Insoluble in ether | |
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2] | |
390.264 | |
390.29 (336.24 Anhydrous) | |
1.533 | |
Barium perchlorate trihydrate |
Safety and Handling
GHS H Statement
H271-H302-H332
P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c
H271-H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BARIUM PERCHLORATE, SOLID
missing translation for 'einecsNumber' : 236-710-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only