missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium peroxide, anhydrous, 84% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Description
Barium peroxide is a specialty product for proteomics research. It is a strong oxidizing agent. It is also used for bleaching in a variety of commercial applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 450°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 84% |
Solubility Information | Insoluble in water |
Formula Weight | 169.34 |
Odor | Odorless |
Sensitivity | Moisture sensitive |
Density | 4.96 g/cm3 |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'