missing translation for 'onlineSavingsMsg'
Learn More
Barium pieces, 2.5cm (1in) & down, 99.2% (metals basis), Sr ∣<0.8%, Thermo Scientific Chemicals Product Code.: 11376607

Barium pieces, 2.5cm (1in) & down, 99.2% (metals basis), Sr ∣<0.8%, Thermo Scientific Chemicals

Product Code. 11376607
50 g
Click to view available options
Quantity:
5 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11376607

Brand: Thermo Scientific Alfa Aesar 000653.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
Synonym bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]

Specifications

Color Silver to Gray
Appearance Silver Gray pieces
Assay Percent Range 99.2% (Metals basis)
Quantity 50 g
Odor Odorless
Form Pieces
Chemical Name or Material Barium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ29I034 Certificate TypeCertificate of Analysis Date05/05/2022 Catalog Number000653.06,000653.18,000653.30
Lot NumberA19P18 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000653.06,000653.18,000653.30
Lot NumberG08N43 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000653.06,000653.18,000653.30
Lot NumberK01Q016 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000653.06,000653.18,000653.30
Lot NumberP27B063 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000653.06,000653.18,000653.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium pieces, 2.5cm (1in) & down, 99.2% (metals basis), Sr ∣<0.8%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.