missing translation for 'onlineSavingsMsg'
Learn More

Barium, plasma standard solution, Specpure™ Ba 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11351459
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Concentration Ba 1000μg/ml
Quantity 50 mL
Molecular Formula Ba(NO3)2 in 5% HNO3
MDL Number MFCD00134031
UN Number UN3264
Physical Form Liquid
Chemical Name or Material Barium, plasma standard solution, Specpure™

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'