Learn More
Barium selenite, 97%, Thermo Scientific™
26.00 € - 100.00 €
Chemical Identifiers
CAS | 13718-59-7 |
---|---|
Molecular Formula | BaO3Se |
Molecular Weight (g/mol) | 264.295 |
MDL Number | MFCD00003446 |
InChI Key | VQKDPABSEYUERS-UHFFFAOYSA-L |
Synonym | barium selenite, selenious acid, barium salt 1:1, barium 2+ selenite, bariumselenit, barium 2+ selenit, acmc-20alqt, bariumselenite, ba.so3e, selenious acid barium salt, seleniousacid,bariumsalt 1:1 |
PubChem CID | 166893 |
IUPAC Name | barium(2+);selenite |
SMILES | [O-][Se](=O)[O-].[Ba+2] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13718-59-7 | |
264.295 | |
VQKDPABSEYUERS-UHFFFAOYSA-L | |
166893 | |
[O-][Se](=O)[O-].[Ba+2] |
BaO3Se | |
MFCD00003446 | |
barium selenite, selenious acid, barium salt 1:1, barium 2+ selenite, bariumselenit, barium 2+ selenit, acmc-20alqt, bariumselenite, ba.so3e, selenious acid barium salt, seleniousacid,bariumsalt 1:1 | |
barium(2+);selenite |
Specifications
13718-59-7 | |
BaSeO3 | |
UN2630 | |
VQKDPABSEYUERS-UHFFFAOYSA-L | |
barium(2+);selenite | |
166893 | |
97% |
BaO3Se | |
MFCD00003446 | |
barium selenite, selenious acid, barium salt 1:1, barium 2+ selenite, bariumselenit, barium 2+ selenit, acmc-20alqt, bariumselenite, ba.so3e, selenious acid barium salt, seleniousacid,bariumsalt 1:1 | |
[O-][Se](=O)[O-].[Ba+2] | |
264.295 | |
264.29 | |
Barium selenite |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES
missing translation for 'einecsNumber' : 237-280-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures