missing translation for 'onlineSavingsMsg'
Learn More

Barium silicon oxide, Thermo Scientific Chemicals

Product Code. 11367098
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Barium silicate films that are suitable for use as transmission media in integrated optical circuits. It is shown that the films, which can be produced with a wide range of refractive indices by suitable selection of the ratio of the target constituents, exhibit low optical attenuation. Hence it is used for integrated optical circuits. It is used in ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13255-26-0
Molecular Formula BaO3Si
Molecular Weight (g/mol) 213.409
MDL Number MFCD00014186
InChI Key HMOQPOVBDRFNIU-UHFFFAOYSA-N
Synonym barium silicate, barium metasilicate, silicic acid, barium salt, barium silicon oxide, barium mesosilicate, barium disilicate, bariumsiliconoxide, ba.so3i, barium meta silicate, silicic acid h2sio3 , barium salt 1:1
PubChem CID 166725
IUPAC Name barium(2+);dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[Ba+2]

Specifications

Color White
Physical Form Powder
Quantity 100 g
Formula Weight 213.4
Chemical Name or Material Barium silicon oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.