missing translation for 'onlineSavingsMsg'
Learn More

Barium sulfate, 98%, extra pure

Product Code. p-4510174 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
1kg
25kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10442031

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Barium sulfate, 98%, BaO4S, CAS Number-7727-43-7, barite, baritop, barium sulfate, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast, 25kg, Passes test, < 0.3%, 97.5 to 100.5%, White, 4.5000g/mL, 231-784-4, 233.4, Extra Pure, 10ppm max., TZCXTZWJZNENPQ-UHFFFAOYSA-L | CAS: 7727-43-7 | BaO4S | 233.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1580.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 97.5 to 100.5%
Linear Formula BaSO4
Merck Index 15,99
Solubility Information Solubility in water: 0.0022g/L (18°C). Other solubilities: practically insoluble in diluted acids, soluble in hot conc. sulfuric acid, insoluble in ethanol
IUPAC Name barium(2+);sulfate
Formula Weight 233.4
Percent Purity 98%
Grade Extra Pure
Loss on Ignition 2.0% (600°C) max.
Packaging Plastic bottle
Specific Gravity 4.5
Density 4.5000g/mL
Chemical Name or Material Barium sulfate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.