missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., 97.5-100.5%, Honeywell Fluka™

Puriss., meets analytical specification of BP, Ph. Eur., 97.5-100.5%
37.55 € - 520.24 €
Chemical Identifiers
CAS | 7727-43-7 |
---|---|
Molecular Formula | BaO4S |
Molecular Weight (g/mol) | 233.38 |
MDL Number | MFCD00003455 |
InChI Key | TZCXTZWJZNENPQ-UHFFFAOYSA-L |
Synonym | barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast |
PubChem CID | 24414 |
IUPAC Name | barium(2+);sulfate |
SMILES | [Ba++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15611050
|
Honeywell-Fluka
11432-100G |
100 g | N/A | N/A | N/A | |||||
15621050
![]() |
Honeywell-Fluka
11432-1KG |
1 kg |
71.28 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15631050
|
Honeywell-Fluka
11432-2.5KG |
2.5 kg | N/A | N/A | N/A | |||||
15641050
![]() |
Honeywell-Fluka
11432-25KG |
25 kg |
520.24 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7727-43-7 | |
233.38 | |
TZCXTZWJZNENPQ-UHFFFAOYSA-L | |
24414 | |
[Ba++].[O-]S([O-])(=O)=O |
BaO4S | |
MFCD00003455 | |
barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast | |
barium(2+);sulfate |
Specifications
7727-43-7 | |
BaSO4 | |
NONH for all modes of transport | |
TZCXTZWJZNENPQ-UHFFFAOYSA-L | |
barium(2+);sulfate | |
24414 | |
97.5 to 100.5% | |
Barium Sulfate, Meets analytical specification of Ph.Eur.BP |
BaO4S | |
MFCD00003455 | |
barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast | |
[Ba++].[O-]S([O-])(=O)=O | |
233.38 | |
233.39g/mol | |
puriss. |
Safety and Handling
missing translation for 'einecsNumber' : 231-784-4
missing translation for 'rtecsNumber' : CR0600000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Barium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., 97.5-100.5%, Honeywell Fluka™