missing translation for 'onlineSavingsMsg'
Learn More

Barium sulfate, precipitated, 98%, Thermo Scientific Chemicals

32.46 € - 266.33 €

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
IUPAC Name barium(2+);sulfate
SMILES [Ba++].[O-]S([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459333
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11768.36
500 g
32.46 €
500g
Please to purchase this item. Need a web account? Register with us today!
11469333
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11768.0E
2500 g
96.18 €
2500g
Please to purchase this item. Need a web account? Register with us today!
11479333
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11768.0C
10,000 g
266.33 €
10000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium sulfate has wide range of applications. It is primarily used in oil well drilling fluid, and as filler in the paint and varnish industry. It is one of the major constituents of the pigment Lithiopone and the white pigment for paints. It improves the resistance against acid and acidity for plastics when employed as filler. Further it is employed as a component in strobe pyrotechnic compositions, and as a coating for photographic papers to improve the effectiveness of the image. Baryta, a thin layer of barium sulfate, is used to make the papers bright for ink-jet printing. In the chemical and pharmaceutical industry, it is used as a catalyst support when selective hydrogenations are required, for e.g. in Rosenmund reduction (Pd on barium sulfate). In colorimetry, barium sulfate is used as a diffuser to measure light sources. In the medical field, barium sulfate suspension is used clinically as a radiocontrast agent for X-ray imaging and other diagnostic procedures. In biomaterials research, it is one of the components utilized in the preparation of acrylic bone cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium sulfate has wide range of applications. It is primarily used in oil well drilling fluid, and as filler in the paint and varnish industry. It is one of the major constituents of the pigment Lithiopone and the white pigment for paints. It improves the resistance against acid and acidity for plastics when employed as filler. Further it is employed as a component in strobe pyrotechnic compositions, and as a coating for photographic papers to improve the effectiveness of the image. Baryta, a thin layer of barium sulfate, is used to make the papers bright for ink-jet printing. In the chemical and pharmaceutical industry, it is used as a catalyst support when selective hydrogenations are required, for e.g. in Rosenmund reduction (Pd on barium sulfate). In colorimetry, barium sulfate is used as a diffuser to measure light sources. In the medical field, barium sulfate suspension is used clinically as a radiocontrast agent for X-ray imaging and other diagnostic procedures. In biomaterials research, it is one of the components utilized in the preparation of acrylic bone cements.

Notes
Incompatible with oxidizing agents, aluminum and phosphorus.
Specifications

Chemical Identifiers

7727-43-7
233.38
TZCXTZWJZNENPQ-UHFFFAOYSA-L
24414
[Ba++].[O-]S([O-])(=O)=O
BaO4S
MFCD00003455
barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
barium(2+);sulfate

Specifications

1580°C (decomposition)
BaO4S
14,994
Soluble in hot concentrated sulphuric acid. Insoluble in water.
[Ba++].[O-]S([O-])(=O)=O
233.38
233.4
Odorless
Barium sulfate, Precipitated
7727-43-7
MFCD00003455
barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
TZCXTZWJZNENPQ-UHFFFAOYSA-L
barium(2+);sulfate
24414
99%
4.5
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-784-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only