missing translation for 'onlineSavingsMsg'
Learn More

Barium Sulfate Precipitated, Extra Pure, SLR, Fisher Chemical™

Product Code. 12618625
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
IUPAC Name barium(2+) sulfate
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1580°C
Color White
Physical Form Powder/Solid
Quantity 2 kg
Formula Weight 233.4g/mol
Packaging Plastic powder jar
Product Identifier
  • Barium sulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'