Learn More
Barium tartrate, 98%, Thermo Scientific Chemicals
22.81 € - 70.55 €
Chemical Identifiers
CAS | 5908-81-6 |
---|---|
Molecular Formula | C4H4BaO6 |
Molecular Weight (g/mol) | 285.397 |
MDL Number | MFCD00054437 |
InChI Key | HQYOWPDUMCBSLQ-UHFFFAOYSA-L |
Synonym | barium tartrate, barium 2+ tartrate, acmc-20alqr, barium 2+ ion tartrate, barium 2+ ; 2,3-dihydroxybutanedioate |
PubChem CID | 6453753 |
IUPAC Name | barium(2+);2,3-dihydroxybutanedioate |
SMILES | C(C(C(=O)[O-])O)(C(=O)[O-])O.[Ba+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424157
|
Thermo Scientific Alfa Aesar
A19604.18 |
50 g |
22.81 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434157
|
Thermo Scientific Alfa Aesar
A19604.30 |
250 g |
70.55 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5908-81-6 | |
285.397 | |
HQYOWPDUMCBSLQ-UHFFFAOYSA-L | |
6453753 | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.[Ba+2] |
C4H4BaO6 | |
MFCD00054437 | |
barium tartrate, barium 2+ tartrate, acmc-20alqr, barium 2+ ion tartrate, barium 2+ ; 2,3-dihydroxybutanedioate | |
barium(2+);2,3-dihydroxybutanedioate |
Specifications
5908-81-6 | |
MFCD00054437 | |
6120238 | |
Very slightly soluble in water | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.[Ba+2] | |
285.397 | |
285.41 | |
Odorless | |
Barium tartrate |
C4H4BaO6 | |
UN1564 | |
barium tartrate, barium 2+ tartrate, acmc-20alqr, barium 2+ ion tartrate, barium 2+ ; 2,3-dihydroxybutanedioate | |
HQYOWPDUMCBSLQ-UHFFFAOYSA-L | |
barium(2+);2,3-dihydroxybutanedioate | |
6453753 | |
98% | |
2.98 |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'einecsNumber' : 227-615-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only