missing translation for 'onlineSavingsMsg'
Learn More

Barium thiosulfate, pure, Thermo Scientific™

Product Code. 15219856
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 35112-53-9
Molecular Formula BaO3S2
Molecular Weight (g/mol) 249.46
InChI Key ONPIOWQPHWNPOQ-UHFFFAOYSA-L
Synonym barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo
PubChem CID 169663
IUPAC Name barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane
SMILES [O-]S(=O)(=S)[O-].[Ba+2]

Specifications

Quantity 2.5 kg
Linear Formula BaS2O3
Merck Index 15,995
Solubility Information Solubility in water: slightly soluble. Other solubilities: insoluble in alcohol
Formula Weight 249.46
Percent Purity 97.5 to 102.5%
Grade Pure
Chemical Name or Material Barium thiosulfate
compliance-icons
Product Identifier
  • Barium thiosulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'