missing translation for 'onlineSavingsMsg'
Learn More

Barium titanate(IV), 99%, Thermo Scientific Chemicals

Product Code. 10694892
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O

Specifications

Melting Point 1625.0°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Powder
Quantity 500 g
Assay Percent Range 0.988 to 1.000 (BaO+ SrO)/TiO2 Mole Ratio
Linear Formula BaTiO3
Merck Index 15, 996
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in alkalies, slightly soluble in diluted acids, soluble in concentrated h2so4 and hf
Formula Weight 233.24
Percent Purity 99%
Loss on Ignition 0.5% max.
Packaging Plastic bottle
Specific Gravity 6.08
Density 6.0800g/mL
Chemical Name or Material Barium titanate(IV), 99%
Show More Show Less
compliance-icons
Product Identifier
  • Barium titanate(IV)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.