Learn More
Barium titanium oxide, 99.7% (metals basis), Thermo Scientific Chemicals
Description
Barium titanium oxide is a dielectric ceramic widely used in multi-layer ceramic capacitors (MLCCs), embedded decoupling capacitors (EDC) and ceramic filters. It is a piezoelectric and ferroelectric component used in microphones and transducers. It is also useful for thermistors and self-regulating electric heating systems. It is doped with small amounts of metals like scandium, yttrium, neodymium, samarium to become a semiconductor. It is used in some types of uncooled sensors for thermal cameras and in electric vehicles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1620°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Merck Index | 14,1000 |
Solubility Information | Slightly soluble in dilute mineral acids. Soluble in concentrated hydrofluoric acid. Insoluble in water. |
Formula Weight | 233.19 |
Percent Purity | 99.7% |
Odor | Odorless |
Density | 5.85 g/mL |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y01K024 | Certificate of Analysis | 16/12/2024 | 088267.22,088267.36 |
R02I078 | Certificate of Analysis | 21/05/2022 | 088267.22,088267.36 |
Z27H015 | Certificate of Analysis | 12/04/2022 | 088267.22,088267.36 |
61400271 | Certificate of Analysis | 08/12/2021 | 088267.22,088267.36 |
H02M27 | Certificate of Analysis | 08/12/2021 | 088267.22,088267.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.