missing translation for 'onlineSavingsMsg'
Learn More
Barium titanium oxide, 99%, Thermo Scientific Chemicals Product Code.: 11331719

Barium titanium oxide, 99%, Thermo Scientific Chemicals

Product Code. 11331719
50 g
Click to view available options
Quantity:
50 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11331719

Brand: Thermo Scientific Alfa Aesar 012348.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ferroelectric ceramics, single crystals, storage devices, dielectric amplifiers

Barium titanium oxide is a dielectric ceramic used for capacitors. It is a piezoelectric material for microphones and other transducers. Finds applications in nonlinear optics and photorefractive applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
Synonym Barium titanate
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O

Specifications

Melting Point 1620°C
Physical Form Powder
Quantity 50 g
Merck Index 14,1000
Solubility Information Insoluble in water, alkalies; Slightly soluble in dilute acids; Soluble in concentrated acids
Formula Weight 233.19
Percent Purity 99%
Odor Odorless
Density 5.85 g/mL
Assay (metals basis)
Chemical Name or Material Barium titanium oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP05L039 Certificate TypeCertificate of Analysis Date07/03/2025 Catalog Number012348.18,012348.A3,012348.36
Lot NumberN12L013 Certificate TypeCertificate of Analysis Date14/02/2025 Catalog Number012348.18,012348.A3,012348.36
Lot NumberN28G048 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number012348.18,012348.A3,012348.36
Lot NumberM20L001 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number012348.18,012348.A3,012348.36
Lot NumberP01J035 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number012348.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium titanium oxide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.