missing translation for 'onlineSavingsMsg'
Learn More

Barium titanium oxide, polymeric precursor, (Oxide ^= 35 wt%), Thermo Scientific Chemicals

Used in piezoelectric devices

102.51 € - 1432.00 €

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394727
View Documents
Thermo Scientific Alfa Aesar
039755.04
2 g
102.51 €
2g
Please to purchase this item. Need a web account? Register with us today!
11374727
View Documents
Thermo Scientific Alfa Aesar
039755.09
10 g
352.76 €
10g
Please to purchase this item. Need a web account? Register with us today!
11384727
View Documents
Thermo Scientific Alfa Aesar
039755.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Barium titanium oxide is used in piezoelectric devices. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12047-27-7
233.19
WNKMTAQXMLAYHX-UHFFFAOYSA-N
[Ba++].[O-][Ti]([O-])=O
BaO3Ti
MFCD00003447
barium(2+) oxotitaniumbis(olate)

Specifications

12047-27-7
BaO3Ti
UN1993
WNKMTAQXMLAYHX-UHFFFAOYSA-N
barium(2+) oxotitaniumbis(olate)
Oxide ≈35 wt%
Viscous Liquid
MFCD00003447
Soluble in alcohols
[Ba++].[O-][Ti]([O-])=O
233.19
Barium titanium oxide, Polymeric precursor
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H332
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'tsca' : Yes

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only