missing translation for 'onlineSavingsMsg'
Learn More

Barium titanium oxide, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 15405965
Click to view available options
Quantity:
2.5 g
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
Synonym Barium titanate
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O

Specifications

Melting Point 1620°C
Color White
Physical Form Powder
Quantity 25 g
Merck Index 14,1000
Formula Weight 233.19
Percent Purity 99.99%
Grade Puratronic™
Odor Odorless
Density 5.85 g/mL
Assay (metals basis)
Chemical Name or Material Barium titanium oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.