Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Brand:  Thermo Scientific™ D5026
 View more versions of this product
Product Code. 11946092
  • 1010.00 € / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pretreated
Nanopure Quad-style system
D5026
Water purification cartridge
One 09-034-5C Macropure™ Pretreatment, 09-050-267 Organicfree, 09-050-276 Ultrapure™ and FS090343 Ultrapure™ cartridges
Cartridge Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Barnstead™ Cartridge Kits > Type I Kit, Organicfree cartridge