missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification Systems

Product Code. p-4531523 Shop All Thermo Scientific Products
Click to view available options
Modules:
3
4
Voltage:
240 V
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12331278

Brand: Thermo Scientific™ D463233

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Produce Type 1 ultrapure water with low operating costs using the Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification system.

Produce Type 1 ultrapure water with low operating costs using Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification Systems. Choose from a three- or four-module system. Gain longer cartridge life and benefit from lower operating costs with large cartridges. This system is Ideal for AA, GS, buffer preps, or as a purified feed water source for analytical instruments or labware washers. It is simple to use and easy to maintain.

AquaPro™ digital on-line analytical measurement platform featuring:

  • Large color graphic display
  • Temperature readout
  • Temperature compensated
  • Calibratable

Features:
  • System achieves resistivities up to 18.2 MΩ-cm and TOC levels below 10ppb.
  • Virgin polypropylene water pathways prevent recontamination.
  • Recirculation pump continuously recirculates water throughout the entire system to maintain water purity and reduce the quantity of wastewater required for rinse-up.
  • System is wall-mountable and completely assembled for easy installation.
  • Quarter-turn quick release canisters allow for easy cartridge replacement.
  • 0.2μm absolute final filter removes bacteria and particulates that could alter your results.
  • Digital resistivity meter is automatically temperature-compensated to 25°C, preventing fluctuating readings due to temperature changes.

Ordering Information:

Optional accessories include Remote Dispenser (D8952), Low Pressure Pump Protector (D2706), and Low Water Level Pump Protector (D0603, D0606). These products, as well as replacement cartridges and consumables, are available separately. For details, contact your sales representative.

Includes:

Wall bracket

Requires:

O-rings included with cartridge pack (sold separately)

Recommended for:

AA, GS, buffer preps, feedwater for analytical instruments or labware washers

Note:

If feeding E-Pure from a storage reservoir, Low Water Pump Protector is recommended to protect pump from damage if reservoir water level falls below system requirements. Order separately (D0603 for 120V, D0606 for 240V).

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, CSA
Conductivity 0.055μS/cm
Bacterial Content <1 CFU/mL
Feed Water Monitoring No
Feedwater Source Tap or Pretreated
Flow Rate Up to 2.5 L/min.
Product Water Type Type 1
Remote Dispenser Optional
Resistivity 18.2 Mohm.cm at 25°C
TOC Monitor with UV Intensity Monitoring No
Total Organic Carbon <10 ppb
Type Water Purification System
Feedwater Pressure 100 psig (6.9 bar)
Modules 3
Particles >0.2µM/mL <1 μM/mL
Dimensions (L x W x H) 7.5 x 31.5 x 28 in. (19 x 80.0 x 71.1 cm)
Length (English) 7.5 in.
Length (Metric) 19 cm
Width (English) 31.5 in.
Width (Metric) 80.0 cm
Height (English) 28 in.
Height (Metric) 71.1 cm
Voltage 240 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.