Learn More
Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application.
200.00 € - 852.24 €
Specifications
For Use With (Equipment) Barnstead Easypure RoDi
View More Specs
Products 6
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10378343
View Documents
Thermo Scientific™
AY1332X1
External Booster Pump This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10317863
View Documents
Thermo Scientific™
AY1332X2
Pre-filtration Accessory Kit
671.00 €
Each
Estimated Shipment: 24-03-2025
to see stock.
Add to basket
10156122
View Documents
Thermo Scientific™
D502132
Start-up Kit
747.00 €
1 set
In Stock
to see stock.
Add to basket
10698333
View Documents
Thermo Scientific™
D502133
Cartridge Kit
409.00 €
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
10433893
View Documents
Thermo Scientific™
D7034
Empty Cartridges
200.00 €
Each
In Stock
to see stock.
Add to basket
10235983
View Documents
Thermo Scientific™
LMX13
Replacement Ultraviolet Lamp
248.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application. Cartridge kit includes pre-filter, ultrapure mixed bed cartridge and Easypure high purity/low TOC cartridge. Other consumables include 0.2 micron final filter, ultraviolet lamp, Thermo Scientific™ Ventgard™ reservoir cap, reverse osmosis membrane, and set of three empty cartridges for cleaning.
Specifications
Specifications
Barnstead Easypure RoDi