Learn More
Thermo Scientific™ Barnstead™ Hose Nipple Cartridges
Purify water up to 75 L/hr using these hose nipple cartridges, ideal for routine lab work including pretreatment.
64.10 € - 237.00 €
Product Code | Brand | Description | Resin Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Resin Type | Price | Quantity & Availability | ||||
15332168
|
Thermo Scientific™
D50220 |
1/2 Size Mixed-Bed | Ultrapure Ion Exchange |
116.00 €
1 unit |
Estimated Shipment: 03-04-2025
Log in to see stock. |
||||
11746299
|
Thermo Scientific™
D8822 |
1/2 Size Mixed-Bed with Oxygen Removal | Ultrapure Mixed Bed with Strong Anion Exchange |
165.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
||||
11810860
|
Thermo Scientific™
D8901 |
Full Size High Capacity | Two Bed Ion Exchange |
155.00 €
Each |
In Stock
Log in to see stock. |
||||
10423865
|
Thermo Scientific™
D8902 |
Full Size Ultrapure | Ultrapure Mixed Bed Ion Exchange |
154.00 €
Each |
In Stock
Log in to see stock. |
||||
12770746
|
Thermo Scientific™
D8903 |
Full Size Oxygen Removal | Strong Anion Exchange |
204.00 €
Each |
In Stock
Log in to see stock. |
||||
10771834
|
Thermo Scientific™
D8904 |
Full Size Organic Removal | Activated Carbon |
136.00 €
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||
10311285
|
Thermo Scientific™
D8905 |
Hose nipple cartridge, Full Size Cation Removal | Cation Exchange |
130.00 €
Each |
In Stock
Log in to see stock. |
||||
15392178
|
Thermo Scientific™
D8908 |
Full Size Macroreticular, Cation Removal | Macroreticular Cation Exchange |
237.00 €
1 unit |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||
12790746
|
Thermo Scientific™
D8911 |
Full Size Ultrapure | Ultrapure Ion Exchange |
172.00 €
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||
11502954
|
Thermo Scientific™
D8921 |
Full Size Pretreatment and Scale Eliminator | Mixed-bed and Strong Cation Exchange, Activated Charcoal |
168.00 €
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
15302188
|
Thermo Scientific™
D8922 |
Full Size Ultrapure with Organic Removal | Mixed-bed Ion Exchange with Activated Carbon |
181.00 €
1 unit |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||
11850850
|
Thermo Scientific™
D8950 |
2/3 Size High Capacity | Two Bed Ion Exchange with Strong Cation Exchange |
145.00 €
Each |
In Stock
Log in to see stock. |
||||
11512954
|
Thermo Scientific™
D8951 |
2/3 Size Mixed-Bed and Organic Removal | Mixed-bed Ion Exchange with Activated Carbon |
141.00 €
Each |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||
10034313
|
Thermo Scientific™
D8900 |
Hose Nipple Cartridge Holder |
64.10 €
Each |
In Stock
Log in to see stock. |
|||||
Description
Purify water volumes up to 75L/hr using cost-efficient Thermo Scientific™ Barnstead™ Hose Nipple Cartridges. Ideal for softening, deionization, and organic and chlorine removal applications, these cartridges are for use with the simple cartridge holder that easily mounts to the wall. Designed for non-pressure applications, these cartridges include 3/8 in. barb hose connections on each end. They provide the ability to see when resin is exhausted in specific cartridges by a color change indicator. Constructed of 100% virgin polypropylene cartridge, the system is customizable and an ideal option for routine laboratory work.
- Ideal for small volume applications
- Same superior quality resins used in B-Pure cartridges
- Constructed of 100% virgin polypropylene cartridge
- Includes 3/8 in. barb hose connections on each end
- D8822, D8950, and D8951 cartridges include tapered straight nipple for 3/8 in. ID tubing
- Color change indicator that identifies when resin is exhausted in specific cartridges