Learn More
Thermo Scientific™ Barnstead™ Mega-Pure™ Accessories
Select from an array of Thermo Scientific™ Barnstead™ Mega-Pure™ Accessories for your Mega-Pure™ Glass Still, including storage bottles and deionization cartridges.
Brand: Thermo Scientific™ D440046
This item is not returnable.
View return policy
Description
Select from an array of Thermo Scientific™ Barnstead™ Mega-Pure™ Accessories for your Mega-Pure Glass Still. Accessories include dual solenoid, deionization cartridges, storage solutions, single cartridge deionizer, dual cartridge deionizer, deionizer accessories and cartridges, storage bottles and automatic collection system.
Specifications
Single Cartridge Deionizer | |
MegaPure Glass Still | |
Single Cartridge Deionizer | |
Single cartridge deionizer removes iNorganic contaminants. Order one deionization cartridge to complete your system. |