Learn More
Thermo Scientific™ Barnstead™ RO System
This product has been discontinued. For additional information or alternate products please contact your sales representative.
Brand: Thermo Scientific™ D12671
Description
This product has been discontinued. For additional information or alternate products please contact your sales representative. The recommended replacement for this product is the Thermo Scientific™ Barnstead™ Pacific™ RO water purification system.
Intelligent Controls:
- Automate operation, monitor system performance and pressure
- Display key data including membrane performance, reservoir level and cartridge status
- Monitor incoming water and membrane pressure and indicate when prefilter must be changed
Simple Operation and Maintenance:
- Delivers multistep purification in a single system
- Includes all required components
Technology Innovations:
- Composite polyamide RO membranes
- Unique MPS that protects membranes from surface scale buildup, eliminates need for softening and extends membrane life
- Automatic flush that cleans RO membrane(s) to help eliminate buildup of scale on membrane and extends membrane life
Compact, Flexible and Quiet:
- Small footprint fits any size lab and features convenient wall- or bench-mounting.
- Wall bracket is included with system.
- Optional Remote Mounting Control Panel enables placement up to 10' (3m) away.
- 12 and 24L/hr. systems include silent-running pump.
- 6L/hr. system uses available in-house line pressure.
Includes:
- Prefilter
- MPS cartridge
- Extruded carbon filter
- RO membrane (two supplied with D12661)
- Wall bracket
Recommended for:
- Buffer/reagent prep
- Glassware washing
- Type 1 water system feed
- General lab use
Ordering Information:
Optional accessories include Remote Control Mounting Panel (AY1265X1) and Hot/Cold Mixing Valve (D7427). These products and replacement consumables are sold separately; contact your Sales Representative for details.
Requires:
Tank, sold separately. Contact your sales representative for details.

Specifications
99% | |
1000 ppm/hr. | |
Monovalent: 90 to 95%; Polyvalent: 95 to 99 | |
90 to 95% | |
Tap | |
Ultrapure (Type 1) | |
Reverse Osmosis System | |
<0.5ppm | |
<1 μM/mL | |
18.2MΩ·cm | |
<5% | |
1°C to 35°C | |
13.5 in. | |
19.5 in. | |
16.9 in. | |
100/240 VAC 50/60 Hz | |
50/60 Hz |
CE, CSA | |
99% | |
99% | |
No | |
6 L/hr. | |
No | |
30 to 100 psig (2 to 6.9 bar) | |
800 ppm | |
Optimum Range: 6.5 to 7.5, Operating Range: 4 to 11 | |
<30ppm | |
34°F to 95°F | |
<1.0 NTU | |
34.3 cm | |
49.5 cm | |
42.9 cm | |
100/240 VAC |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.