Learn More
Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables
Optimize your Thermo Scientific™ Barnstead™ RO and TII water purification system with these accessories and consumables, including filters, cartridges and dispensers.
Brand: Thermo Scientific™ D502114
This item is not returnable.
View return policy
Description
Optimize your water purification system with Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables. For use with Barnstead RO and TII water purification systems, these products include filters and pre-filters, cartridges, overflow cutoff float and pump interlock cable, as well as dispensers and printer.
Optimize your water purification system with Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables. For use with Barnstead RO and TII water purification systems, these products include filters and pre-filters, cartridges, overflow cutoff float and pump interlock cable, as well as dispensers and printer.Specifications
MPS Cartridge | |
Barnstead RO, Barnstead TII | |
Cartridge, MPS |